2-Chloro-5-nitropyrimidin-4-amine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Amino-2-chloro-5-nitropyrimidine
H64478.06
H64478.06EA
357
EUR
H64478.06
2-Chloro-5-nitropyrimidin-4-amine 97%
2-Chloro-5-nitropyrimidin-4-amine
Purity: 97%
CAS No.: 1920-66-7
Molecular Formula: C4H3ClN4O2
Formula Weight: 174.54
Physical Form: Solid
Melting Point: 221-226°
Beilstein Reference: 156953
MDL No.: MFCD00127771
Formel:
C₄H₃ClN₄O₂ Molecular Weight: 174.544 g/mol Schmelzpunkt: 221…226 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00127771 CAS-Nummer: 1920-66-7 EINECS: 217-648-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...