5-(Trifluoromethyl)picolinonitrile 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Cyano-5-(trifluoromethyl)pyridine
H64486.03
H64486.06
H64486.03EA
34.7
EUR
H64486.03
H64486.06
5-(Trifluoromethyl)picolinonitrile 98%
5-(Trifluoromethyl)picolinonitrile
Purity: 98%
CAS No.: 95727-86-9
Molecular Formula: C7H3F3N2
Formula Weight: 172.11
Physical Form: Solid
Beilstein Reference: 13339826
UN No.: UN3439
MDL No.: MFCD01001119
Formel:
C₇H₃F₃N₂ Molecular Weight: 172.11 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01001119 CAS-Nummer: 95727-86-9 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...