N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 98%
Lieferant: Thermo Fisher Scientific
H64502.MD
H64502.03
H64502.06
H64502.MDEA
49.5
EUR
H64502.MD
H64502.03
H64502.06
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine 98%
N,N'-Di-1-naphthyl-N,N'-diphenylbenzidine
Purity: 98%
CAS No.: 123847-85-8
Molecular Formula: C44H32N2
Formula Weight: 588.75
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 279-283°
Beilstein Reference: 8022378
MDL No.: MFCD03093246
Formel:
C₄₄H₃₂N₂ Schmelzpunkt: 279…283 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03093246 CAS-Nummer: 123847-85-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...