2-(4-Fluorophenyl)-1H-benzimidazole 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(4-Fluorophenyl)benzimidazole, 2-(4-Fluorophenyl)-1H-benzo[d]imidazole
H64531.03
H64531.03EA
305
EUR
H64531.03
2-(4-Fluorophenyl)-1H-benzimidazole 95%
2-(4-Fluorophenyl)-1H-benzimidazole
Purity: 95%
CAS No.: 324-27-6
Molecular Formula: C13H9FN2
Formula Weight: 212.23
Physical Form: Solid
MDL No.: MFCD00224358
Formel:
C₁₃H₉FN₂ Molecular Weight: 212,227 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00224358 CAS-Nummer: 324-27-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...