5-Bromo-3-chloro-2-fluoropyridine 95%
Lieferant: Thermo Fisher Scientific
H64547.06
H64547.06EA
349
EUR
H64547.06
5-Bromo-3-chloro-2-fluoropyridine 95%
5-Bromo-3-chloro-2-fluoropyridine
Purity: 96%
CAS No.: 38185-56-7
Molecular Formula: C5H2BrClFN
Formula Weight: 210.43
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 60°
Beilstein Reference: 1563886
MDL No.: MFCD09834365
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...