Tetrahydropyran-4-methanol 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H64568.03
H64568.06
H64568.03EA
72.8
EUR
H64568.03
H64568.06
Tetrahydropyran-4-methanol 98%
Tetrahydropyran-4-methanol
Purity: 98%
CAS No.: 14774-37-9
Molecular Formula: C6H12O2
Formula Weight: 116.16
Flash Point: 61°(142°F)
Physical Form: Liquid
Refractive Index: 1.4600
Boiling Point: 111-115°/15mm
Beilstein Reference: 103069
MDL No.: MFCD00457804
Formel:
C₆H₁₂O₂ Siedepunkt: 111…115 °C (15 mmHg) Flammpunkt: 61 °C (142 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00457804 CAS-Nummer: 14774-37-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...