3-Fluoro-4-methoxypyridine 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64603.06
H64603.03
H64603.06EA
417
EUR
H64603.06
H64603.03
3-Fluoro-4-methoxypyridine 97%
3-Fluoro-4-methoxypyridine
Purity: 98%
CAS No.: 1060805-03-9
Molecular Formula: C6H6FNO
Formula Weight: 127.12
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 19561531
MDL No.: MFCD13185605
Formel:
C₆H₆FNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185605 CAS-Nummer: 1060805-03-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...