2-Amino-4-chlor-3-nitropyridin 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Chloro-3-nitropyridin-2-amine
H64678.03
H64678.06
H64678.03EA
94.1
EUR
H64678.03
H64678.06
2-Amino-4-chlor-3-nitropyridin 95%
2-Amino-4-chloro-3-nitropyridine
Purity: 95%
CAS No.: 6980-08-1
Molecular Formula: C5H4ClN3O2
Formula Weight: 173.56
Beilstein Reference: 391939
MDL No.: MFCD04116097
Formel:
C₅H₄ClN₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04116097 CAS-Nummer: 6980-08-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...