3-Fluor-2-methoxy-6-methylpyridin 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Fluoro-2-methoxy-6-methylpyridine, 3-Fluoro-2-methoxy-6-picoline
H64683.06
H64683.06EA
132
EUR
H64683.06
3-Fluor-2-methoxy-6-methylpyridin 97%
3-Fluoro-2-methox-6-methylypyridine
Purity: 97%
CAS No.: 375368-80-2
Molecular Formula: C7H8FNO
Formula Weight: 141.15
Physical Form: Liquid
Appearance: Colourless to pale yellow
MDL No.: MFCD03095083
Formel:
C₇H₈FNO Molecular Weight: 141,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095083 CAS-Nummer: 375368-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...