1H-Pyrimidin-4-on 98%

Lieferant: Thermo Scientific

Synonyme: 4-Hydroxypyrimidin

H64690.06
H64690.06EA 131 EUR
H64690.06
1H-Pyrimidin-4-on 98%
1H-Pyrimidin-4-on

Purity: 98% CAS No.: 4562-27-0 Molecular Formula: C4H4N2O Formula Weight: 96.09 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 163-169° lit. Beilstein Reference: 741972 MDL No.: MFCD00006664

Formel: C₄H₄N₂O
Molecular Weight: 96,09 g/mol
Schmelzpunkt: 163…169 °C (Lit.)
MDL: MFCD00006664
CAS-Nummer: 51953-17-4
EINECS: 224-932-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR