tert-Butyl hydrogen succinate 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64706.03
H64706.06
H64706.03EA
40.6
EUR
H64706.03
H64706.06
tert-Butyl hydrogen succinate 97%
tert-Butyl hydrogen succinate
Purity: 97%
CAS No.: 15026-17-2
Molecular Formula: C8H14O4
Formula Weight: 174.20
Flash Point:
Formel:
C₈H₁₄O₄ Schmelzpunkt: 51…54 °C (Lit.) Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00273441 CAS-Nummer: 15026-17-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...