4-(Trifluormethyl)-1H-imidazol 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)imidazole
H64711.03
H64711.06
H64711.03EA
40
EUR
H64711.03
H64711.06
4-(Trifluormethyl)-1H-imidazol 98%
4-(Trifluoromethyl)-1H-imidazole
Purity: 98%
CAS No.: 33468-69-8
Molecular Formula: C4H3F3N2
Formula Weight: 136.08
Physical Form: Solid
Melting Point: 148-150°
MDL No.: MFCD08458860
Formel:
C₄H₃F₃N₂ Schmelzpunkt: 148…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08458860 CAS-Nummer: 33468-69-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...