6-Chloro-5-fluoropicolinic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Chloro-3-fluoropyridine-6-carboxylic acid
H64734.03
H64734.06
H64734.03EA
53.1
EUR
H64734.03
H64734.06
6-Chloro-5-fluoropicolinic acid 98%
6-Chloro-5-fluoropicolinic acid
Purity: 98%
CAS No.: 860296-24-8
Molecular Formula: C6H3ClFNO2
Formula Weight: 175.54
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 10063182
MDL No.: MFCD13185819
Formel:
C₆H₃ClFNO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD13185819 CAS-Nummer: 860296-24-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...