2-Amino-9,9-dimethylfluorene 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 9,9-Dimethyl-9H-fluoren-2-amine

H64771.06
H64771.06EA 219 EUR
H64771.06
2-Amino-9,9-dimethylfluorene 98%
2-Amino-9,9-dimethylfluorene

Purity: 98% CAS No.: 108714-73-4 Molecular Formula: C15H15N Formula Weight: 209.29 Physical Form: Powder Appearance: Off-white Beilstein Reference: 15506865 MDL No.: MFCD09953790

Formel: C₁₅H₁₅N
Molecular Weight: 209,29 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09953790
CAS-Nummer: 108714-73-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR