8-Brom-1-octen 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H64830.03
H64830.06
H64830.03EA
46.3
EUR
H64830.03
H64830.06
8-Brom-1-octen 97%
8-Brom-1-octen
Purity: 97%
CAS No.: 2695-48-9
Molecular Formula: C8H15Br
Formula Weight: 191.11
Flash Point: 78°(172°F)
Physical Form: Liquid
Density: 1.139
Refractive Index: 1.4670
Appearance: Pale yelllow
Boiling Point: 92°/24mm
Beilstein Reference: 2234481
MDL No.: MFCD00000275
Formel:
C₈H₁₅Br Siedepunkt: 92 °C (24 mmHg) Dichte: 1,139 Flammpunkt: 78 °C (172 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000275 CAS-Nummer: 2695-48-9 EINECS: 220-268-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...