4-Chloro-2-(trifluoromethyl)pyridine 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
H64833.06
H64833.06EA
238
EUR
H64833.06
4-Chloro-2-(trifluoromethyl)pyridine 98%
4-Chloro-2-(trifluoromethyl)pyridine
Purity: 98%
CAS No.: 131748-14-6
Molecular Formula: C6H3ClF3N
Formula Weight: 181.54
Physical Form: Liquid
Appearance: Colourless
UN No.: UN2810
MDL No.: MFCD07368047
Formel:
C₆H₃ClF₃N Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368047 CAS-Nummer: 131748-14-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...