3-(9H-Carbazol-9-yl)phenylboronic acid 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-(9-Carbazolyl)benzeneboronic acid, 3-(9H-Carbazol-9-yl)benzeneboronic Acid, (3-(9H-Carbazol-9-yl)phenyl)boronic acid
H64898.03
H64898.06
H64898.03EA
56
EUR
H64898.03
H64898.06
3-(9H-Carbazol-9-yl)phenylboronic acid 98%
3-(9H-Carbazol-9-yl)phenylboronic acid
Purity: 98%
CAS No.: 864377-33-3
Molecular Formula: C18H14BNO2
Formula Weight: 287.13
Storage Temperature: 2°C to 8°C
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 15530380
MDL No.: MFCD18072506
Formel:
C₁₈H₁₄BNO₂ Molecular Weight: 287.13 g/mol |
MDL:
MFCD18072506 CAS-Nummer: 864377-33-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...