Copper(I)-2-thiophenecarboxylate

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (2-Thiophenecarboxylato)copper(I), Copper(I) thiophene-2-carboxylate, 2-Thiophenecarboxylic Acid Copper(I) Salt, CuTC

H64950.03 H64950.06
H64950.03EA 92.6 EUR
H64950.03 H64950.06
Copper(I)-2-thiophenecarboxylate
Copper(I)-2-thiophenecarboxylate

CAS No.: 68986-76-5 Molecular Formula: C5H3CuO2S Formula Weight: 190.68 Physical Form: Powder Sensitivity: Air and Light Sensitive Appearance: Red to brown MDL No.: MFCD02183524

Formel: C₅H₃CuO₂S
Molecular Weight: 190.68 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02183524
CAS-Nummer: 68986-76-5

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR