Ethyl-2-aminonicotinat 98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Ethyl 2-aminonicotinate
H64957.06
H64957.14
H64957.22
H64957.06EA
43.3
EUR
H64957.06
H64957.14
H64957.22
Ethyl-2-aminonicotinat 98%
Ethyl-2-aminonicotinat
Purity: 98%
CAS No.: 13362-26-0
Molecular Formula: C8H10N2O2
Formula Weight: 166.18
Physical Form: Solid
Melting Point: 90-96° lit.
Beilstein Reference: 124680
MDL No.: MFCD01569452
Formel:
C₈H₁₀N₂O₂ Molecular Weight: 166.18 g/mol Schmelzpunkt: 90…96 °C (Lit.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01569452 CAS-Nummer: 13362-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...