D-Histidine methyl ester dihydrochloride 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(R)-Methyl 2-amino-3-(1H-imidazol-4-yl)propanoate dihydrochloride, H-D-His-OMe.2HCl
H65096.03
H65096.06
H65096.14
H65096.03EA
40.2
EUR
H65096.03
H65096.06
H65096.14
D-Histidine methyl ester dihydrochloride 95%
D-Histidine methyl ester dihydrochloride
Purity: 95%
CAS No.: 4467-54-3
Molecular Formula: C7H11N3O2.2HCl
Formula Weight: 242.10
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Hygroscopic
Appearance: Off-white to white
Beilstein Reference: 6491805
Optical Rotation: -9°(c=2 in water)
MDL No.: MFCD00066137
Formel:
C₇H₁₁N₃O₂ Molecular Weight: 242.10 g/mol |
MDL:
MFCD00066137 CAS-Nummer: 4467-54-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...