Cloflucarban 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
1-(4-Chlorophenyl)-3-[4-chloro-3-(trifluoromethyl)phenyl]urea
H65397.MD
H65397.MDEA
71.5
EUR
H65397.MD
Cloflucarban 97%
Cloflucarban
Purity: 97%
CAS No.: 369-77-7
Molecular Formula: C14H9Cl2F3N2O
Formula Weight: 349.13
Physical Form: Solid
MDL No.: MFCD00867294
Formel:
C₁₄H₉Cl₂F₃N₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00867294 CAS-Nummer: 369-77-7 EINECS: 206-724-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...