3-Iod-1-methyl-1H-pyrazol 97%
Lieferant: Thermo Scientific
|
Gefahr
|
H66356.03
H66356.03EA
180
EUR
H66356.03
3-Iod-1-methyl-1H-pyrazol 97%
3-Iodo-1-methyl-1H-pyrazole
Purity: 97%
CAS No.: 92525-10-5
Molecular Formula: C4H5IN2
Formula Weight: 208.00
Flash Point: 110°(230°F)
Physical Form: Liquid
Density: 1.936
Refractive Index: 1.5880
Sensitivity: Light Sensitive
Appearance: Brown
MDL No.: MFCD08435890
Formel:
C₄H₅IN₂ Molecular Weight: 208,00 g/mol Dichte: 1,936 Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD08435890 CAS-Nummer: 92525-10-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...