PPDT2FBT (PCE9.3), Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Fisher Scientific
H66399.MC
H66399.MD
H66399.ME
H66399.MCEA
489
EUR
H66399.MC
H66399.MD
H66399.ME
PPDT2FBT (PCE9.3), Reagenz für organische Photovoltaik (OVP)
PPDT2FBT (PCE9.3)
Formel:
(C₅₂H₇₂F₂N₂O₂S₃)n |
CAS-Nummer:
1620673-07-5 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,78 ± 0,02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...