Dde-Lys(Fmoc)-OH 98%

Lieferant: Thermo Scientific

Synonyme: Nα-1-(4,4-Dimethyl-2,6-dioxocyclohex-1-ylidene)ethyl-Nepsilon-Fmoc-L-lysine, DDE-Lys(Fmoc)-OH

H66446.03
H66446.03EA 246 EUR
H66446.03
Dde-Lys(Fmoc)-OH 98%
Dde-Lys(Fmoc)-OH

Purity: 98% CAS No.: 156648-40-7 Molecular Formula: C31H36N2O6 Formula Weight: 532.64 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: Off-white MDL No.: MFCD00467662

Formel: C₃₁H₃₆N₂O₆
Molecular Weight: 532,637 g/mol
MDL: MFCD00467662
CAS-Nummer: 156648-40-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR