Y5, Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Fisher Scientific
H66460.MC
H66460.MD
H66460.ME
H66460.MCEA
673
EUR
H66460.MC
H66460.MD
H66460.ME
Y5, Reagenz für organische Photovoltaik (OVP)
Y5
Formel:
C₈₂H₉₀N₈O₂S₅ |
CAS-Nummer:
2304444-48-0 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,41 ± 0,02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...