Fmoc-Glu-OBzl 95%

Lieferant: Thermo Scientific

Synonyme: Fmoc-Glu-Obzl, N-Fmoc-L-glutamic acid 1-benzyl ester

H66551.06
H66551.06EA 90.5 EUR
H66551.06
Fmoc-Glu-OBzl 95%
Fmoc-Glu-OBzl

Purity: 95% CAS No.: 122350-52-1 Molecular Formula: C27H25NO6 Formula Weight: 459.50 Storage Temperature: -30°C to -10°C Physical Form: Powder Appearance: White Optical Rotation: -16°(c=1 in methanol) MDL No.: MFCD00080272

Formel: C₂₇H₂₅NO₆
Molecular Weight: 459,498 g/mol
MDL: MFCD00080272
CAS-Nummer: 122350-52-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR