IEICO-4Cl, Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Fisher Scientific
H66752.MC
H66752.MD
H66752.ME
H66752.MCEA
612
EUR
H66752.MC
H66752.MD
H66752.ME
IEICO-4Cl, Reagenz für organische Photovoltaik (OVP)
IEICO-4Cl
Formel:
C₁₁₄H₁₁₄Cl₄N₄O₄S₄ |
CAS-Nummer:
2240998-88-1 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | 1,26 ± 0,02 eV |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...