Benzyl viologen dichloride 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
H66836.03
H66836.06
H66836.03EA
96.7
EUR
H66836.03
H66836.06
Benzyl viologen dichloride 97%
Benzyl viologen dichloride
Purity: 97%
CAS No.: 1102-19-8
Molecular Formula: C24H22Cl2N2
Formula Weight: 409.35
Physical Form: Solid
Appearance: White to off-white
Melting Point: 260° dec.
Beilstein Reference: 3811585
MDL No.: MFCD00011991
Formel:
C₂₄H₂₂Cl₂N₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011991 CAS-Nummer: 1102-19-8 EINECS: 214-158-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...