PBDB-T-2Cl, Reagenz für organische Photovoltaik (OVP)
Lieferant: Thermo Fisher Scientific
H66867.MC
H66867.MD
H66867.ME
H66867.MCEA
673
EUR
H66867.MC
H66867.MD
H66867.ME
PBDB-T-2Cl, Reagenz für organische Photovoltaik (OVP)
PBDB-T-2Cl
Formel:
(C₆₈H₇₆Cl₂O₂S₈)n |
CAS-Nummer:
2239295-71-5 |
Spezifikation Testergebnisse
Appearance (Color) | Black |
Form | Flake or threadlike |
Bandgap energy | Tbd |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...