Hämatoxylin, Supelco®
Lieferant: Merck
H9627-5X100G
H9627-5X100GEA
1650
EUR
H9627-5X100G
Hämatoxylin, Supelco®
Hämatoxylin
Hematoxylin is an organic stain which chelates with metal ions and binds to DNA and RNA. It is used for staining nucleus of the cells.[1] Hematoxylin along with eosin stain is used in the diagnosis of invasion stage in tumors.
Formel:
C₁₆H₁₄O₆ |
MDL:
MFCD00078111 CAS-Nummer: 517-28-2 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...