Rotor CC38/Ti with ''Connect Assist'' and ceramic shaft ***************************************************** Coaxial cylinder with ‘’Connect Assist’’, ceramic shaft and integral solvent trap ring for use with cup CCB43 for TM-XX-C48. Material: Titan 1 * 1 ST
Lieferant: HAAKE
Rotor CC38/Ti with ''Connect Assist'' and ceramic shaft ***************************************************** Coaxial cylinder with ‘’Connect Assist’’, ceramic shaft and integral solvent trap ring for use with cup CCB43 for TM-XX-C48. Material: Titan 1 * 1 ST