Powder pillows, Chloride 2 indicator
Lieferant: Hach
104399
105766
HACH104399EA
49.9
EUR
HACH104399
HACH105766
Powder pillows, Chloride 2 indicator
Umwelt-Prüfkits
Powder reagent in sealed package used as indicator for the argentometric titration of chloride.
Bitte beachten Note: Product contains potsassium chromate. Please read MSDS prior to usage.