Powder pillows, Chloride 2 indicator
Lieferant: Hach
104399 105766
HACH104399EA 49.9 EUR
HACH104399 HACH105766
Powder pillows, Chloride 2 indicator
Umwelt-Prüfkits
Powder reagent in sealed package used as indicator for the argentometric titration of chloride.
Bitte beachten Note: Product contains potsassium chromate. Please read MSDS prior to usage.