Silikonöl

Lieferant: Hach

126936
HACH126936EA 20.6 EUR
HACH126936
Silikonöl
Silikonöl

Multiple uses, including preparation of turbidity sample cells.

Dichte: 0,9…1.1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00132673
CAS-Nummer: 68083-14-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR