Membrane disc filters
Lieferant: Hach
1353001
HACH1353001EA 260 EUR
HACH1353001
Membrane disc filters
Filter Membranfilter
These GN-metrical membrane filters are ideal for testing recreational water, wastewater and drinking water. They meets U.S. Environmental Protection Agency (USEPA) and American Society for Testing and Materials (ASTM) specifications to ensure complete retention and maximum recovery of bacteria.
  • Convenient and pre-sterilised
  • Gridded design