These GN-metrical membrane filters are ideal for testing recreational water, wastewater and drinking water. They meets U.S. Environmental Protection Agency (USEPA) and American Society for Testing and Materials (ASTM) specifications to ensure complete retention and maximum recovery of bacteria.