Potassium iodide-iodate standard solution, 0,0125 N
Lieferant: Hach
1400153
HACH1400153EA 65.6 EUR
HACH1400153
Potassium iodide-iodate standard solution, 0,0125 N
Standards Umweltanalyse-Standards
Used for determination of sulphite by burette iodometric titration.