Solution, cobalt chloride, for spectrophotometer
Lieferant: Hach
1422249
HACH1422249EA 88.2 EUR
HACH1422249
Solution, cobalt chloride, for spectrophotometer
Standards Umweltanalyse-Standards
Cobalt chloride solution for spectrophotometer linearisation.