Demineraliser bottle
Lieferant: Hach
1429900
HACH1429900EA 31 EUR
HACH1429900
Demineraliser bottle
Flaschen Kunststoffflaschen
This narrow mouth polyethylene demineraliser bottle is used to deionize small volumes of water.
  • Flip spout cap
  • Contain monobed resin inside the bottle