Bromphenolblau-Lösung Säure-Base-Indikator
Lieferant: Hach
Synonyme:
3,3',5,5'-Tetrabromphenolsulfonphthalein, 4,4'-(3H-2,1-Benzoxathiol-3-yliden)bis[2,6-dibromphenol]-S,S-dioxid
1455232
HACH1455232EA
23.3
EUR
HACH1455232
Bromphenolblau-Lösung Säure-Base-Indikator
Bromphenolblau
Bromphenol blue pH indicator solutions are used to determine acidity and alkalinity of water by titration method.
- Visual transition interval: pH 3,0 (yellow) - pH 4,6 (blue)
Formel:
C₁₉H₁₀Br₄O₅S Molecular Weight: 669,97 g/mol |
MDL:
MFCD00005875 CAS-Nummer: 115-39-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...