Arsenic standard solution
Lieferant: Hach
1457142
HACH1457142EA 29.3 EUR
HACH1457142
Arsenic standard solution
Standards Kationischer Standards Arsen-Standards
Standard solution for accuracy checks in arsenic determinations.