Chloride standard solution
Lieferant: Hach
18349
HACH18349EA 65.4 EUR
HACH18349
Chloride standard solution
Standards Anionische Standards Chlorid-Standards
For accuracy checks in chloride determinations. Also suitable for standardising conductivity apparatus.