Phenolphthalein 0,1% in wässriger Lösung

Lieferant: Hach

Synonyme: 3,3-Bis(4-hydroxyphenyl)phthalid, 3,3-Bis(4-hydroxyphenyl)-1(3H)-isobenzofuranon

189736
HACH189736EA 16.1 EUR
HACH189736
Phenolphthalein 0,1% in wässriger Lösung
Phenolphthalein
Formel: C₂₀H₁₄O₄
Molecular Weight: 318.33 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00005913
CAS-Nummer: 77-09-8
EINECS: 201-004-7
Merck Index: 14,07243
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8d81b539a32ba7c40a473e6afbb8e0dae525d637&vid=91bf141a7ea82baf5df563f73967884d2a4b5d79&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR