Eisen(III)chlorid-Lösung in Schwefelsäure

Lieferant: Hach
Gefahr

Synonyme: Eisentrichlorid

204253
HACH204253EA 41 EUR
HACH204253
Eisen(III)chlorid-Lösung in Schwefelsäure
Eisen(III)chlorid

Ferric Chloride-Sulfuric Acid Solution for determination of volatile acids by the esterification method.

Formel: FeCl₃
Molecular Weight: 162,21 g/mol
MDL: MFCD00011005
CAS-Nummer: 7705-08-0
UN: 3264
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR