Nitrogen-nitrate standard solution
Lieferant: Hach
2415132 1426010 194749 204649
HACH2415132EA 17.7 EUR
HACH2415132 705-2010 705-2025 HACH204649
Nitrogen-nitrate standard solution
Standards Umweltanalyse-Standards
Standard solutions for accuracy checks in nitrate nitrogen determinations and also for meter calibrations/standardisation.