Formaldehyd 36 - 38% aqueous solution
Lieferant: Hach
|
Gefahr
|
Synonyme:
Methanal, Formalin, Formol, Methylaldehyd
205932
HACH205932EA
16.5
EUR
HACH205932
Formaldehyd 36 - 38% aqueous solution
Formaldehyd
Formel:
CH₂O Molecular Weight: 30.03 g/mol Siedepunkt: -19.3 Schmelzpunkt: -92 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003274 CAS-Nummer: 50-00-0 EINECS: 200-001-8 UN: 2209 ADR: 8 III Merck Index: 13,04259 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...