Thymolblau

Lieferant: Hach

Synonyme: 4-[3-(4-Hydroxy-2-methyl-5-propan-2-ylphenyl)-1,1-dioxobenzo[c]oxathiol-3-yl]-5-methyl-2-propan-2-ylphenol, Thymolsulfonphthalein

2067299
HACH2067299EA 44.1 EUR
HACH2067299
Thymolblau
Thymolblau
Formel: C₂₇H₃₀O₅S
Molecular Weight: 466,6 g/mol
Schmelzpunkt: 221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005869
CAS-Nummer: 76-61-9
EINECS: 200-973-3
Merck Index: 12,09541

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR