Erlenmeyer flasks, polymethylpentene
Lieferant: Hach
2089846
HACH2089846EA 34.7 EUR
HACH2089846
Erlenmeyer flasks, polymethylpentene
Kolben Erlenmeyerkolben
Ideal for titrations, preparation and storage of media.
  • Chemical resistance Strength
  • Autoclavable
Polymethylpentene flask with polypropylene screwcap features clarity (Do not engage threads during cycle).