Brush, camel hair
Lieferant: Hach
2103200
HACH2103200EA 4.3 EUR
HACH2103200
Brush, camel hair
Bürsten
For cleaning delicate instruments or to ensure complete transfer of samples when weighing.
  • Soft, brown hair is cemented into a metal holder
  • Hairs are cut to the same length: 14 mm (0,5")
  • Can be attached to the Hach micro spatula (HACH1225600)