Neßler-Reagenz

Lieferant: Hach

Synonyme: Potassium mercuric Iodide, Potassium tetraiodomercurate, Nota 1, Mercury (II) potassium iodide, Potassium tetraiodomercurate – Potassium hydroxide solution

2119432 2119449
HACH2119432EA 32.1 EUR
HACH2119432 458-0044
Neßler-Reagenz
Neßler-Reagenz

The reagent is used for ammonia nitrogen determination in waste water by the Nessler method

MDL: MFCD00049656
CAS-Nummer: 7783-33-7

Bestellen

Spezifikation Testergebnisse

Range 0.02-2.50 mg/L NH3-N


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR