1-(2-Pyridylazo)-2-Naphthol (PAN) -Lösung 0,1% Indikator

Lieferant: Hach
Gefahr

Synonyme: PAN

2122432 2122426 2122449
HACH2122432EA 39.3 EUR
HACH2122432 HACH2122426 HACH2122449
1-(2-Pyridylazo)-2-Naphthol (PAN) -Lösung 0,1% Indikator
1-(2-Pyridylazo)-2-Naphthol (PAN)

For determination of low level manganese by the PAN method.

Formel: C₁₅H₁₁N₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD00004071
CAS-Nummer: 85-85-8
UN: 3082
ADR: 9,III

Bestellen

Spezifikation Testergebnisse

Method 8149
Method Name PAN
Parameter Manganese
Platform Solution
Range 0.006 - 0.700 mg/L Mn


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR