Hardness reagent, UniVer® 3
Lieferant: Hach
21320H
HACH21320HEA 31.4 EUR
HACH21320H
Hardness reagent, UniVer® 3
Umwelt-Prüfkits
This combined indicator and buffer reagent is used for determination of total hardness by titration methods.